Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Acros Organics

Potassium Chloride, 99+%, ACS Reagent

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 424090050
HSN 28273990
Price (INR)
35,578.00
Each
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Chlorate and Nitrate (as NO3)=<0.003 %
Bromide (Br)=<0.01 %
Barium (Ba)Passes test (Limit about 0.001%)
Iodide (I)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only