Potassium sulfate, 99+%, ACS reagent
Potassium sulfate, 99+%, ACS reagent
Potassium sulfate, 99+%, ACS reagent
Potassium sulfate, 99+%, ACS reagent
Potassium sulfate, 99+%, ACS reagent
Acros Organics

Potassium sulfate, 99+%, ACS reagent

CAS: 7778-80-5 | K2O4S | 174.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number 424220010
also known as 42422
HSN 28332990
Price (INR)
9,776.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
9,776.00
Each
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=99.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.01 %
Nitrogen compounds (as N)=<5 ppm
pH5.5 to 8.5 (5% soln. at 25°C)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only