RadEye PRD4 and PRD-ER4 Personal Radiation Detector
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Actual product may vary
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Thermo Scientific™
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
The Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors provide highly sensitive radiation detection capability with little to no nuisance alarms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDetector TypeDescription
4250691731 Low Dose Rate Detector, 1 High Dose Rate DetectorPRD-ER4 Vehicle Kit
4250671261 Low Dose Rate DetectorPRD4 Pocket-sized Personal Detector
4250671271 Low Dose Rate Detector, 1 High Dose Rate DetectorPRD-ER4 Pocket-sized Personal Detector
4250691601 Low Dose Rate DetectorPRD4 Charger Package
4250691611 Low Dose Rate DetectorPRD4 Charger/BTLE Package
4250691621 Low Dose Rate DetectorPRD4 Kit
4250691631 Low Dose Rate DetectorPRD4 Vehicle Kit
4250691701 Low Dose Rate Detector, 1 High Dose Rate DetectorPRD-ER4 Charger Package
4250691711 Low Dose Rate Detector, 1 High Dose Rate DetectorPRD-ER4 Charger/BTLE Package
4250691721 Low Dose Rate Detector, 1 High Dose Rate DetectorPRD-ER4 Kit
Catalog number 425069173
Price (INR)
-
Request A Quote
Detector Type:
1 Low Dose Rate Detector, 1 High Dose Rate Detector
Description:
PRD-ER4 Vehicle Kit
Achieve highly sensitive radiation detection with few nuisance alarms with Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors. RadEye PRD4 and PRD-ER4 radiation detectors save users valuable time adjudicating alarms while increasing scanning throughput. Using Natural Background Rejection (NBR) Technology, the RadEye PRD4 and PRD-ER4 radiation detectors ignore fluctuations in naturally occurring radiation (NORM) while analyzing the energy imbalance of artificial radiation. Additionally, these radiation detectors identify the discovered material in a manner configurable for your operation or user skills.
The RadEye PRD4 and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD4 provides an additional layer of detection critical to determining the level of radiation threat.
Efficiency in the field
  • Larger display
  • Neutron indication capability
  • 25% improved sensitivity over previous model
  • Linear high dose rate detector
Easy information, smarter decisions
  • No retraining or relearning for infrequent users
  • Get results automatically, without the need to press buttons
  • Quickly guides you through next steps after an alarm
  • Simple 4 button design
  • Comprehensive data neatly organized and presented on screen
Rugged and practical
  • Can be worn in holster or standard service belt
  • Small and lightweight
  • Long battery life
  • Drop resistant to 1.5m
  • IP65
  • Can be operated in extreme temperatures
View our online training here.
Specifications
DescriptionPRD-ER4 Vehicle Kit
Detector Type1 Low Dose Rate Detector, 1 High Dose Rate Detector
IncludesRadEye PRD-ER4, Lutetium Test Adapter, Vehicle charging kit, Bluetooth adapter, Software package RadEye.exe, Holster and spare AAA rechargeable batteries
Product TypePersonal Radiation Detector
Unit SizeEach