FHT 192 Ionization Chambers
Actual product may vary
Thermo Scientific™

FHT 192 Ionization Chambers

Thermo Scientific™ FHT 192 is a wide-range ionization chamber for high-precision ambient equivalent dose rate measurements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
425354050FHT 192 (Built into carrying case with external connector)
4253540FHT 192
Catalog number 425354050
Price (INR)
-
Description:
FHT 192 (Built into carrying case with external connector)

Thermo Scientific™ FHT 192 is a wide-range ionization chamber for high-precision ambient equivalent dose rate measurements. Ideal for connecting to either the FHT 6020 area monitor or the FH 40 G survey meter for measurements at high energy accelerators or X-ray sources.

The FHT 192 Ionization Chamber is based on the famous PTB-approved medium-pressure chamber FHT 191 N, which is used as a reference standard detector by many organizations.
  • Chamber geometry design offers both excellent angular dependence (-45° to +90°) and energy dependence
  • Chamber is equipped with Styrofoam protection in order to minimize temperature change-induced currents
  • 30keV to 7MeV energy range
  • 100nSv/h to 1Sv/h dose rate range
  • 0.75 sphere angular response

  • Integrated preamplifier fits to: FHT 6020 area monitors; FH 40 G survey meter; and SVG2 military Radiac
  • High dose option with remote preamplifier

Compatible with:

FHT 6020 area monitors, FH 40 G survey meter, and SVG2

Specifications
Angular Acceptance-45° <F <+90° (270° solid angle)
DescriptionFHT 192 (Built into carrying case with external connector)
Dose Range10 µR/h to 100 R/h
Energy Range30 keV to 7 MeV(± 30%)
Gamma250 fA/µSvh-1
GasNoble Gas - Nitrogen - Mixture
OverloadUp to 50 Sv/h
Pressure7000 hPa
Radiation TypeGamma
TypeIonization Chamber
Volume (Metric)4.25 L
Weight (English)7.1 lb.
Weight (Metric)3.25 kg
Unit SizeEach