2,3-Diaminophenol, 97%
2,3-Diaminophenol, 97%
2,3-Diaminophenol, 97%
Acros Organics

2,3-Diaminophenol, 97%

CAS: 59649-56-8 | C6H8N2O | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 425680050
also known as 42568
HSN 29222990
Price (INR)
46,467.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
46,467.00
Each
Chemical Identifiers
CAS59649-56-8
IUPAC Name2,3-diaminophenol
Molecular FormulaC6H8N2O
InChI KeyPCAXITAPTVOLGL-UHFFFAOYSA-N
SMILESNC1=C(N)C(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Melting point161°C to 165°C
Appearance (Color)Light brown to dark brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only