2-Methyl-3-buten-1-ol, 98%
2-Methyl-3-buten-1-ol, 98%
2-Methyl-3-buten-1-ol, 98%
Acros Organics

2-Methyl-3-buten-1-ol, 98%

CAS: 4516-90-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 425690050
also known as 42569
HSN 29052900
Price (INR)
54,799.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
54,799.00
Each
Chemical Identifiers
CAS4516-90-9
IUPAC Name2-methylbut-3-en-1-ol
Molecular FormulaC5H10O
InChI KeyNVGOATMUHKIQQG-UHFFFAOYNA-N
SMILESCC(CO)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4240 to 1.4300 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only