Methyl undecanoate, 97%
Methyl undecanoate, 97%
Methyl undecanoate, 97%
Acros Organics

Methyl undecanoate, 97%

CAS: 1731-86-8 | C12H24O2 | 200.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 426580050
also known as 42658
HSN 29159090
Price (INR)
6,770.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,770.00
Each
Chemical Identifiers
CAS1731-86-8
IUPAC Namemethyl undecanoate
Molecular FormulaC12H24O2
InChI KeyXPQPWPZFBULGKT-UHFFFAOYSA-N
SMILESCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only