3-Bromo-2-methylpropene, 97%
3-Bromo-2-methylpropene, 97%
3-Bromo-2-methylpropene, 97%
Acros Organics

3-Bromo-2-methylpropene, 97%

CAS: 1458-98-6 | C4H7Br | 135.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 427650050
also known as 42765
HSN 29033919
Price (INR)
9,445.00
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,445.00
Each
Chemical Identifiers
CAS1458-98-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
GC>=96.0 %
StabilizerApprox 500 ppm hydroquinone
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only