(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]
(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]
(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]
Acros Organics

(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]

CAS: 150529-93-4 | C21H22N2O2 | 334.42 g/mol
Have Questions?
Catalog NumberQuantity
428050010
also known as 42805
1 g
Catalog number 428050010
also known as 42805
HSN 32049000
Price (INR)
31,933.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
31,933.00
Each
Chemical Identifiers
CAS150529-93-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
GC>=96.0 %
Specific optical rotation+150° to +170° (20°C, 589 nm) (c=1%, ethanol)
Appearance (Form)viscous liquid or solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only