Diazolidinyl urea, 95%
Diazolidinyl urea, 95%
Diazolidinyl urea, 95%
Acros Organics

Diazolidinyl urea, 95%

CAS: 78491-02-8 | C8H14N4O7 | 278.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number 428582500
also known as 42858
HSN 32049000
Price (INR)
39,271.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,271.00
Each
Chemical Identifiers
CAS78491-02-8
IUPAC Name1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
Molecular FormulaC8H14N4O7
InChI KeySOROIESOUPGGFO-UHFFFAOYNA-N
SMILESOCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<3 %
Elemental Analysis19.0 to 21.5 % N
Solubility(50 mg/ml in water) Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only