2-Bromo-3'-hydroxyacetophenone, 96%
2-Bromo-3'-hydroxyacetophenone, 96%
2-Bromo-3'-hydroxyacetophenone, 96%
Acros Organics

2-Bromo-3'-hydroxyacetophenone, 96%

CAS: 2491-37-4 | C8H7BrO2 | 215.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428620010
also known as 42862
1 gGlass bottle
Catalog number 428620010
also known as 42862
HSN 29145000
Price (INR)
14,061.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,061.00
Each
Chemical Identifiers
CAS2491-37-4
IUPAC Name2-bromo-1-(3-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H7BrO2
InChI KeyIEPSGFQQGKPTPM-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Appearance (Color)Brown
HPLC>=95 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only