N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
Acros Organics

N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™

CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
429021000
also known as 42902
100 mLAcroSeal™ Glass Bottle
Catalog number 429021000
also known as 42902
HSN 29251900
Price (INR)
5,989.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
5,989.00
Each
Chemical Identifiers
CAS538-75-0
IUPAC NameN,N'-dicyclohexylmethanediimine
Molecular FormulaC13H22N2
InChI KeyQOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILESC1CCC(CC1)N=C=NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO40.95 to 1.10 M
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only