Amberlite™ IRN-78, OH-form, ion-exchange resin
Amberlite™ IRN-78, OH-form, ion-exchange resin
Amberlite™ IRN-78, OH-form, ion-exchange resin
Acros Organics

Amberlite™ IRN-78, OH-form, ion-exchange resin

CAS: 11128-95-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 429295000
HSN 98020000
Price (INR)
10,250.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
10,250.00
Each
Chemical Identifiers
CAS11128-95-3
MDL NumberMFCD00145822
SpecificationsSpecification SheetSpecification Sheet
Uniformity coefficient=<1.20
Total volume capacity>=1.10 eq/l (OH) as is
Appearance (Form)Spherical beads
Moisture holding capacity54.0 to 60.0 % (OH)
Harmonic mean size0.580 to 0.680 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only