3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
Acros Organics

3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran

CAS: 862129-81-5 | C11H19BO2S | 226.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 429610010
also known as 42961
HSN 29319090
Price (INR)
29,163.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
29,163.00
Each
Chemical Identifiers
CAS862129-81-5
IUPAC Name2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC11H19BO2S
InChI KeyQZVRTORVCKGPPY-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CCSCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
Infrared spectrumConforms
Appearance (Color)Orange to brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only