Methyl 2-aminopyridine-4-carboxylate, 98%
Methyl 2-aminopyridine-4-carboxylate, 98%
Methyl 2-aminopyridine-4-carboxylate, 98%
Acros Organics

Methyl 2-aminopyridine-4-carboxylate, 98%

CAS: 6937-03-7 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 430060010
also known as 43006
HSN 29333919
Price (INR)
7,977.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,977.00
Each
Chemical Identifiers
CAS6937-03-7
IUPAC Namemethyl 2-aminopyridine-4-carboxylate
Molecular FormulaC7H8N2O2
InChI KeySVWWNEYBEFASMP-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=NC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Melting point144°C to 148°C
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only