Wash Solution I, RNA Purification
Invitrogen™

Wash Solution I, RNA Purification

This Wash Solution is for use with the Total RNA Chemistry System.Total RNA Chemistry makes possible the rapid isolation ofRead more
Have Questions?
Catalog NumberQuantity
43058911000 mL
Catalog number 4305891
Price (INR)
38,445.33
Each
Quantity:
1000 mL
Price (INR)
38,445.33
Each
This Wash Solution is for use with the Total RNA Chemistry System.

Total RNA Chemistry makes possible the rapid isolation of total RNA from various samples, including tissue-cultured cells, whole blood, and peripheral blood mononuclear cells (PBMCs). The protocol is designed for use with the 6100 Nucleic Acid PrepStation and 6700 Automated Nucleic Acid Workstation.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)6100 Nucleic Acid PrepStation, 6700 Automated Nucleic Acid Workstation
Quantity1000 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.