Single-Channel Dual 384-well Temperature Verification Kit for PCR 9700 System, 0.02 mL
Have Questions?
Catalog Number
Quantity
4308354
1 kit
Catalog number 4308354
Price (INR)
-
Quantity:
1 kit
The Temperature Verification Kits and Probes are engineered and manufactured specifically for Applied Biosystems to validate temperature calibration and uniformity in GeneAmp™ PCR and the Veriti™ Thermal Cycler instrument systems. This product is for use with the Dual 384-Well GeneAmp™ PCR System 9700.
• Self-contained digital thermometer and battery operated for ease of use • Interchangeable Platinum Resistant probes
Complete Offering To expand the use of an existing Temperature Verification Kits, the individual platinum resistant probes are now available in 0.02 mL, 0.1mL, 0.2 mL, for purchase.
More information can also be found at Alpha Technics (Formerly Alpha Sensors, Inc., Eutechnics Division) (www.alphatechnics.com). Calibration standards are traceable to NIST (US National Institute of Standards and Technology). The kit contains one platinum resistant probe, hand held thermometer, mineral oil, cotton swabs and carrying case.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)GeneAmp 9700
FormatKit
Quantity1 kit
Shipping ConditionRoom Temperature
Product TypeTemperature Verification Kit
Unit SizeEach
Contents & Storage
Store at Room Temperature
Frequently asked questions (FAQs)
Do you offer a temperature verification service to verify my PCR/qPCR instrument performance?
Yes, we offer an on-site temperature verification service with additional features such as ramp rate testing. Please find more information regarding the service below: https://www.thermofisher.com/us/en/home/products-and-services/services/instrument-qualification-services/compliance-and-validation/on-site-temperature-verification.html