Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™
Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™
Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™
Acros Organics

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™

CAS: 22722-98-1 | C6H16AlNaO4 | 202.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 430911000
also known as 43091
HSN 29319090
Price (INR)
8,498.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (INR)
8,498.00
Each
Chemical Identifiers
CAS22722-98-1
IUPAC Namealumanylium sodium bis(2-methoxyethan-1-olate)
Molecular FormulaC6H16AlNaO4
InChI KeyXJIQVZMZXHEYOY-UHFFFAOYSA-N
SMILES[Na+].[AlH2+].COCC[O-].COCC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Appearance (Color)Clear to slightly hazy colorless to amber
Assay68.0 to 72.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only