tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate, 97%
tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate, 97%
tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate, 97%
Acros Organics

tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate, 97%

CAS: 782501-25-1 | C10H18ClNO4S | 283.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 430990010
also known as 43099
HSN 29333990
Price (INR)
42,778.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
42,778.00
Each
Chemical Identifiers
CAS782501-25-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis40.84 to 43.81 % (C)
Appearance (Color)White to pale yellow
Melting point82°C to 87°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only