1,3-Benzothiazole-6-carboxaldehyde, 97%
1,3-Benzothiazole-6-carboxaldehyde, 97%
1,3-Benzothiazole-6-carboxaldehyde, 97%
Acros Organics

1,3-Benzothiazole-6-carboxaldehyde, 97%

CAS: 19989-67-4 | C8H5NOS | 163.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 431150050
also known as 43115
HSN 29342000
Price (INR)
91,798.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
91,798.00
Each
Chemical Identifiers
CAS19989-67-4
IUPAC Name1,3-benzothiazole-6-carbaldehyde
Molecular FormulaC8H5NOS
InChI KeyAVSFPLJXSHRMHM-UHFFFAOYSA-N
SMILESO=CC1=CC=C2N=CSC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point90°C to 96°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to yellow
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only