MicroAmp™ Optical Film Compression Pad
Applied Biosystems™
MicroAmp™ Optical Film Compression Pad
Applied Biosystems™ MicroAmp™ Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler andRead more
Have Questions?
Catalog NumberQuantity
43126395 Ea.
Catalog number 4312639
Price (INR)
8,860.00
Each
Quantity:
5 Ea.
Price (INR)
8,860.00
Each
Applied Biosystems™ MicroAmp™ Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films.
Note: See user's manual or package insert for limited label license, and trademark information.
Specifications
For Use With (Application)PCR
For Use With (Equipment)2720 Thermal Cycler, 3100 Genetic Analyzer, 3700 Genetic Analyzer, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp 9700, Veriti Thermal Cycler, 2720 Thermal Cycler
Product TypeCompression Pad
Quantity5 Ea.
Shipping ConditionRoom Temperature
ColorOptical
Plate Compatibility96-well Plate
Product LineMicroAmp
Unit SizeEach
Contents & Storage
5 Optical Cover Compression Pads
Frequently asked questions (FAQs)
Can I use an MicroAmp Optical Cover Compression Pad on an Optical Fast 96-Well plate?
No. we do not recommend placing compression pads on the Fast 96-well plate or the StepOnePlus, ViiA 7, 7300, 7500, 7900HT, or 7500 Fast Real-Time PCR Systems.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?
No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How many times can a MicroAmp Optical Film Compression Pad (Cat. No. 4312639) be used?
With proper usage, the compression pad should maintain performance for a minimum of 20 PCR runs. It is important to examine the pad before each run, especially after more than 20 uses, as deteriorated compression pads may lead to evaporation and failed reactions.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.