Product Image
Applied Biosystems™
dCbz, 0.2 μmol ABI 3900 HT Columns
Have Questions?
Catalog NumberQuantity
4316676200 Columns
Catalog number 4316676
Price (INR)
0.01
Each
Quantity:
200 Columns
Price (INR)
0.01
Each
The 0.2 μmol d(Cbz) ABI 3900 HT columns are fully assembled, low-cost columns that produce high-quality oligonucleotides. These supports are filled with polystryene, a material which has several advantages over the more conventional, controlled pore glass (CPG) filled columns.
• Proprietary design and manufacturing methods ensure consistency and high quality to give you optimal synthesis quality.
• Hydrophobic, polystyrene-filled columns sustain an anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis.
• Polystyrene support allows synthesis of oligonucleotides of a variety of lengths, giving you flexibility in high-throughput oligonucleotide synthesis.
Designed for Performance
The greater hydrophobicity of polystyrene results in a higher yield of full-length product. Side reactions, such as extraneous chain growth, are minimized by the inert nature of this support surface.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeOligonucleotide Synthesis Column
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormLiquid
High-throughput CompatibilityHigh-throughput Compatible
MatrixPolystyrene
Prep Scale200 nmol
Product TypeOligonucleotide Synthesis Column
Quantity200 Columns
Sample TypeOligos
Shipping ConditionRoom Temperature
TargetNucleic Acids
FormatColumn
Unit SizeEach
Contents & Storage
Columns may be stored in a dry environment at room temperature.