Product Image
Applied Biosystems™
TaqMan™ RNase P Control Reagents Kit
The RNase P gene is a single-copy gene that encodes the RNA moiety for the RNase P enzyme. Applied Biosystems™Read more
Have Questions?
Catalog NumberQuantity
43168441,000 reactions
Catalog number 4316844
Price (INR)
82,975.00
Each
Quantity:
1,000 reactions
Price (INR)
82,975.00
Each
The RNase P gene is a single-copy gene that encodes the RNA moiety for the RNase P enzyme. Applied Biosystems™ TaqMan™ RNase P Control Reagents were designed with limiting primer concentrations to be used as the endogenous reference in multiplex reactions. Multiplex PCR is the use of more than one probe and primer pair in the same tube. This kit is designed to perform a 5' nuclease assay with TaqMan™ Universal PCR Master Mix using the provided human genomic DNA as template.
• Pre-designed primers and TaqMan™ probe eliminate assay design
• Easy-to-use primer and TaqMan™ probe mix minimizes assay set-up time
• Designed with limiting primer concentrations to eliminate assay optimization
• Rapid assay development guidelines can minimize optimization time
Comprehensive Guidelines Included
Applied Biosystems™ has developed a comprehensive set of guidelines to ensure success when using Applied Biosystems™ Real-Time PCR reagents and instrumentation. These guidelines are simple and easy to follow. Many traditional variables, such as magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, greatly reducing assay development time.
Normalization Built-In
All Applied Biosystems™ Real-Time PCR reagent kits contain a passive internal reference to normalize non-PCR related fluorescence fluctuations. Normalizing with a passive internal reference minimizes well-to-well variability that can result from a variety of causes, such as pipetting errors and sample evaporation. The passive reference is also essential for accurate results when multiple probes (with different reporter dyes) are combined in a single tube.
Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibration TypePassive Internal Reference Standards
Control TemplateHuman Genomic DNA
FormatTube
Internal Probe ModificationTAMRA™ Quencher (3'), VIC™ (5')
No. of Reactions1000 Reactions
Product LineTaqMan
Product TypeReal Time PCR Reagents
Quantity1,000 reactions
Shipping ConditionWet Ice
SpeciesHuman
Sufficient For1000 Reactions
TargetRNase P
GC-Rich PCR PerformanceLow
Label or DyeVIC
PCR MethodMultiplex qPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Control Primer-Probe Set(s) & Template(s)