Biotin-HRP (biotinylated peroxidase)
Invitrogen™

Biotin-HRP (biotinylated peroxidase)

Biotin-HRP (biotinylated peroxidase)
Have Questions?
Catalog NumberQuantity
4320405 mg
Catalog number 432040
Price (INR)
-
Quantity:
5 mg
Reactivity: Reacts with streptavidin and avidin conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity5 mg
Shipping ConditionWet Ice
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator at 2°C to 8°C