Product Image
Applied Biosystems™

Syringe Adaptor, 3100/3730

This is a Syringe Adaptor, 3100/3730 - to be used with a 20 mL plastic syringe for block cleaning.
Have Questions?
Catalog NumberQuantity
43229281 adaptor
Catalog number 4322928
Price (INR)
-
Quantity:
1 adaptor

This is a Syringe Adaptor, 3100/3730 - to be used with a 20 mL plastic syringe for block cleaning. For the 3730/3730xl, it applies to v1.0 syringe systems only.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)3100 Genetic Analyzer, 3730 DNA Analyzer
Product TypeSyringe Adaptor
Quantity1 adaptor
Shipping ConditionRoom Temperature
Volume (Metric) Well20 mL
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Store at room temperature.