Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%
Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%
Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%
Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%
Acros Organics

Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%

CAS: 42303-42-4 | C6H11NO2·ClH | 165.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 432510250
also known as 43251
HSN 29224990
Price (INR)
63,937.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS42303-42-4
IUPAC Nameethyl 1-aminocyclopropane-1-carboxylate hydrochloride
Molecular FormulaC6H12ClNO2
InChI KeyXFNUTZWASODOQK-UHFFFAOYSA-N
SMILESCl.CCOC(=O)C1(N)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis42.41 to 45.48 % C
Appearance (Form)Crystalline powder
Melting point112°C to 118°C
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only