Product Image
Applied Biosystems™

Serial Adapter, G4PORT G4 (for the 310 Genetic Analyzer)

Have Questions?
Catalog NumberQuantity
43262451 Unit
Catalog number 4326245
Price (INR)
-
Quantity:
1 Unit
This is a spare part for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeSerial Adapter
Quantity1 Unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.