2-Methyl-1,3-benzothiazol-5-amine, 95%
2-Methyl-1,3-benzothiazol-5-amine, 95%
2-Methyl-1,3-benzothiazol-5-amine, 95%
Acros Organics

2-Methyl-1,3-benzothiazol-5-amine, 95%

CAS: 13382-43-9 | C8H8N2S | 164.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432760010
also known as 43276
HSN 29342000
Price (INR)
7,007.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,007.00
Each
Chemical Identifiers
CAS13382-43-9
IUPAC Name2-methyl-1,3-benzothiazol-5-amine
Molecular FormulaC8H8N2S
InChI KeyGPWQHYMVUZYWIK-UHFFFAOYSA-N
SMILESCC1=NC2=CC(N)=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point100°C to 105°C
HPLC>=94.0 %
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only