TaqMan™ GMO Detection Kits User's Guide
Product Image
Applied Biosystems™
TaqMan™ GMO Detection Kits User's Guide
The TaqMan™ Genetically Modified Organism (GMO) Detection Kits User's Guide provides all information necessary for the use of TaqMan™ GMORead more
Have Questions?
Catalog NumberQuantity
43276871 protocol
Catalog number 4327687
Price (INR)
-
Request A Quote
Quantity:
1 protocol
The TaqMan™ Genetically Modified Organism (GMO) Detection Kits User's Guide provides all information necessary for the use of TaqMan™ GMO Assays.
The GMO detection kits contain reagents needed to detect with high confidence the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods and their ingredients.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Kit TypeGMO Detection Kit
Product LineTaqMan™
Quantity1 protocol
Shipping ConditionRoom Temperature
Unit SizeEach