Product Image
Applied Biosystems™

7900HT TaqMan™ Array Upgrade

The 7900HT TaqMan™ Array Upgrade enables you to run the TaqMan™ Arrays – a microfluidic card – on your AppliedRead more
Have Questions?
Catalog NumberQuantity
43290121 kit
Catalog number 4329012
Price (INR)
-
Quantity:
1 kit
The 7900HT TaqMan™ Array Upgrade enables you to run the TaqMan™ Arrays – a microfluidic card – on your Applied Biosystems 7900HT Fast Real-Time PCR System. This upgrade gives you access to an efficient, easy-to-use tool that eliminates labor-intensive pipetting steps and enables high throughput gene expression results using the gold-standard 5' nuclease TaqMan™ Assay technology.

• Convenience – includes everything you need to run the 7900HT TaqMan™ Array on your 7900HT system
• Success – highly trained service engineers will install and test your 7900HT TaqMan™ Array Upgrade to get you up and running quickly
• High throughput – provides access to 384-well format without liquid-handling robotics

This upgrade to your 7900HT system provides everything you need to run the TaqMan™ Array.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
Format384-well Microfluidic Card
Product LineTaqMan
Quantity1 kit
Reaction SpeedFast
Block FormatInterchangeable
Product TypeArray Upgrade
Unit SizeEach
Contents & Storage
This kit contains a thermal cycling block, a microfluidic card sealer, centrifuge buckets with adapters, and a chemistry installation kit.

Frequently asked questions (FAQs)

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.