Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Acros Organics

Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

CAS: 13125-66-1 | C7H15BrMg | 203.41 g/mol
Have Questions?
Catalog NumberQuantityPackaging
433031000
also known as 43303
100 mLAcroSeal™ Glass Bottle
Catalog number 433031000
also known as 43303
HSN 29319090
Price (INR)
36,359.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
36,359.00
Each
Chemical Identifiers
CAS13125-66-1
IUPAC Namebromo(heptyl)magnesium
Molecular FormulaC7H15BrMg
InChI KeyLDKGBIJDNGSVIH-UHFFFAOYSA-M
SMILESCCCCCCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.95 to 1.10 M
Appearance (Color)Brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only