Product Image
Applied Biosystems™
GeneScan™ Installation Standard DS-33 , [6FAM™, VIC™, NED™, PET™ dyes]
Using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347.
Have Questions?
Catalog NumberQuantity
43303971 Kit
Catalog number 4330397
Price (INR)
77,745.00
Each
Quantity:
1 Kit
Price (INR)
77,745.00
Each
The DS-33 GeneScan™ Installation Standards using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in Human DNA. The kit also contains a tube of GeneScan™ 500 LIZ™ Size Standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeFAM, NED, PET, VIC
Quantity1 Kit
Shipping ConditionDry Ice
For Use With (Equipment)3730 DNA Analyzer, 3730xl DNA Analyzer
Product LineFAM, NED PET, VIC
TypeInstallation Standard
Unit SizeEach
Contents & Storage
This kit contains 4 tubes of fluorescently labeled pooled PCR products and 1 tube of GeneScan 500 LIZ Size Standard.
Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 500 LIZ Size Standard tube at 2-8°C.