Product Image
Applied Biosystems™

Plate Base (heat seal), 96 well

Have Questions?
Catalog NumberQuantity
43348754 Each
Catalog number 4334875
Price (INR)
-
Quantity:
4 Each
This is a 96-well Plate Base (Heat Seal) for use with Applied Biosystems™ 3730 and 3730xl DNA Analyzers.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3730xl DNA Analyzer, 3730 DNA Analyzer
Plate Compatibility96-well plate, 96-Well Plates
Product TypePlate Base
Quantity4 Each
Shipping ConditionRoom Temperature
Format96-well Plate
Unit SizeEach
Contents & Storage
Each package contains 4 units.