Product Image
Applied Biosystems™

Autosampler Installation Kit (Septa), 96-well plate

The 96-Well Plate Autosampler Kit (for use with septa) for 3730 and 3730xl DNA Analyzer. This kit includes the plateRead more
Have Questions?
Catalog NumberQuantity
43361311 Ea.
Catalog number 4336131
Price (INR)
-
Quantity:
1 Ea.
The 96-Well Plate Autosampler Kit (for use with septa) for 3730 and 3730xl DNA Analyzer. This kit includes the plate base, retainer, septa and twenty 96-well microplates. This system enables plates to be attached to the instrument autosampler.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3730 DNA Analyzer
Product TypeAutosampler Installation Kit
Quantity1 Ea.
Format96-well Plate
Unit SizeEach
Contents & Storage
This kit includes the plate base, retainer, septa and twenty 96-well microplates.