Product Image
Applied Biosystems™

StockMarks™ for Horses 17-Plex Genotyping Kit, equine, with protocol

This PCR-based genotyping system helps validate the parentage of horses quickly and accurately.
Have Questions?
Catalog NumberQuantity
43364071 Kit
Catalog number 4336407
Price (INR)
-
Quantity:
1 Kit

This PCR-based genotyping system helps validate the parentage of horses quickly and accurately. The StockMarks™ for Horses Equine 17-plex Genotyping Kit includes 17 microsatellite markers recommended by the International Society for Animal Genetics (ISAG), co-amplified in a single tube.

This kit includes Protocol.

  • PCR process provides greater accuracy than serological methods.
  • ISAG-recommended, standardized loci ensure accurate testing.
  • Proprietary primer purification process ensures optimal data quality.

Accurate with All Sample Types
All StockMarks™ kits amplify microsatellite markers by means of PCR and fluorescent dye-labeled primers. PCR is more accurate than serological typing and requires less sample, is easier to standardize and automate, and is successful with all types of samples, including blood, semen, and hair. Additionally, when fluorescent dye is used to label PCR primers, multiple amplifications can be performed in a single reaction.

Microsatellite-Based Kits Provide Accurate Parentage Testing
The StockMarks™ Horse Genotyping System comprises the StockMarks™ for Horses Equine Genotyping Kit. This kit both confirm or exclude parentage, using the highly accurate and informative polymorphic short tandem repeats (STRs), also known as microsatellites, which are evenly distributed across the genome.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection MethodCapillary Electrophoresis
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, GeneAmp 9600, GeneAmp 9700
Hot StartBuilt-In Hot Start
Label or DyeFAM, NED, PET, VIC
PCR MethodMultiplex PCR
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineStockMarks
Product TypeGenotyping Kit
Quantity1 Kit
Sample TypeDNA
SpeciesHorse⁄Equine, Horse
TargetEquine Genotyping and Parentage Testing
Target LociASB2, LEX3, ASB17, HMS3, AHT5, CA425, VHL20, HMS6, HTG10, AHT4, HTG6, HTG7, HMS1, HMS2, ASB23, HTG4, HMS57
TechniqueSTR (Short Tandem Repeat) Analysis
FormatKit
Unit SizeEach
Contents & Storage
This kit includes the dye-labeled primer mix, dNTP solution, required buffer, bovine control DNA, AmpliTaq Gold™ DNA polymerase, and protocol. Enough reagents for 100 assays are supplied.

Store reagents at -15 to -25°C.