9-Fluorenylmethyl carbamate, 99%
9-Fluorenylmethyl carbamate, 99%
9-Fluorenylmethyl carbamate, 99%
Acros Organics

9-Fluorenylmethyl carbamate, 99%

CAS: 84418-43-9 | C15H13NO2 | 239.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 434170010
also known as 43417
HSN 29241900
Price (INR)
12,481.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,481.00
Each
Chemical Identifiers
CAS84418-43-9
IUPAC Name(9H-fluoren-9-yl)methyl carbamate
Molecular FormulaC15H13NO2
InChI KeyZZOKVYOCRSMTSS-UHFFFAOYSA-N
SMILESNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to off-white
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only