Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%
Acros Organics

Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%

CAS: 4160-61-6 | C7H10O3S | 174.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 434290050
also known as 43429
HSN 32049000
Price (INR)
84,081.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
84,081.00
Each
Chemical Identifiers
CAS4160-61-6
IUPAC Namemethyl 4-oxothiane-3-carboxylate
Molecular FormulaC7H10O3S
InChI KeyMCUXKFHPGMEIIW-UHFFFAOYNA-N
SMILESCOC(=O)C1CSCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or solid
GC>=94.0 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only