5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%
5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%
5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%
Acros Organics

5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%

CAS: 894086-00-1 | C32H35N4P | 506.62 g/mol
Have Questions?
Catalog NumberQuantityPackaging
435160010
also known as 43516
1 gGlass bottle
Catalog number 435160010
also known as 43516
HSN 29319090
Price (INR)
34,205.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,205.00
Each
Chemical Identifiers
CAS894086-00-1
IUPAC Name5-(di-tert-butylphosphanyl)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole
Molecular FormulaC32H35N4P
InChI KeyPTXJGGGNGMPMBG-UHFFFAOYSA-N
SMILESCC(C)(C)P(C1=CC=NN1C1=C(N(N=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
NMR>=96.0 %
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only