(S)-2-Amino-7-hydroxytetralin, 95%
(S)-2-Amino-7-hydroxytetralin, 95%
(S)-2-Amino-7-hydroxytetralin, 95%
Acros Organics

(S)-2-Amino-7-hydroxytetralin, 95%

CAS: 85951-60-6 | C10H13NO | 163.22 g/mol
Have Questions?
Catalog NumberQuantity
435590050
also known as 43559
5 g
Catalog number 435590050
also known as 43559
HSN 29222990
Price (INR)
130,122.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
130,122.00
Each
Chemical Identifiers
CAS85951-60-6
IUPAC Name7-amino-5,6,7,8-tetrahydronaphthalen-2-ol
Molecular FormulaC10H13NO
InChI KeyVIYAPIMIOKKYNF-UHFFFAOYNA-N
SMILESNC1CCC2=C(C1)C=C(O)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-85° to -95° (20°C, 589 nm) (c=0.5, MeOH)
Appearance (Form)Crystalline Powder or crystals
Melting point144°C to 151°C
Appearance (Color)White to grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only