Plates and Modules with Covalent Binding Surfaces
Actual product may vary
Thermo Scientific™

Plates and Modules with Covalent Binding Surfaces

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Have Questions?
Catalog NumberColorVolume (Metric) WorkingProduct TypeSurface Treatment
436009Clear50 μLSolid plate 384-WellAmine
Catalog number 436009
Price (INR)
-
Color:
Clear
Volume (Metric) Working:
50 μL
Product Type:
Solid plate 384-Well
Surface Treatment:
Amine

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Binding TypeCovalent
Detection MethodColorimetric
FormatStandard
LidWithout Lid
No. per Case30
No. per Pack5
Packaging30 Cases
Plate BlockingUnspecified
Product TypeSolid plate 384-Well
Quantity30/Cs.
Target MoleculePeptides
Total Volume (Metric) Well120 μL
Volume (Metric) Well120 μL
Volume (Metric) Working50 μL
Array16 x 24
ColorClear
MaterialPolystyrene
No. of Wells384
SterilityNon-sterile
Surface TreatmentAmine
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 30

Frequently asked questions (FAQs)

Which type of Immobilizer Amino plates and strips should I use for the different detection methods?

We recommend using transparent polystyrene plates and strips for colorimetric assays, white opaque polystyrene plates and strips for bio and chemi-luminescence assays, and black opaque polystyrene plates and strips for fluorescence assays.

What advantages do your covalent binding surfaces offer over passive surfaces?

While adsorption of biomolecules to our passive surfaces is more electrostatic in nature, our CovaLink and Immobilizer Amino surfaces present an option for true covalent bonding of molecules with specific free residues. CovaLink possesses a secondary amine structure, which will covalently bond to biomolecules with a free carboxyl or phosphate group. This allows for a much more robust and specific coating, which will often tolerate more rigorous washing steps and eliminate the need for blocking.