CHAPS, 98%, Thermo Scientific Chemicals
CHAPS, 98%, Thermo Scientific Chemicals
CHAPS, 98%, Thermo Scientific Chemicals
Acros Organics

CHAPS, 98%, Thermo Scientific Chemicals

CHAPS, 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, CAS # 75621-03-3, is a non-denaturing zwitterionic detergent that can be used for solubilizing membrane proteins.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 436550050
HSN 29241900
Price (INR)
31,341.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
31,341.00
Each
Chemical Identifiers
CAS75621-03-3
IUPAC Name3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylazaniumyl)propane-1-sulfonate
Molecular FormulaC32H58N2O7S
InChI KeyUMCMPZBLKLEWAF-BCTGSCMUSA-N
SMILESC[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water=<2 % (K.F.)
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • CHAPS (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate) is a zwitterionic non-denaturing detergent
  • It is suitable for applications which require protection of native protein conformation

Applications

  • CHAPS can be used for the solubilization and purification of membrane proteins
  • It is also used in conjunction with nonionic detergents like Triton X-100
  • It can be used as a non-denaturing zwitterionic detergent for the study of membrane chemistry
RUO – Research Use Only