Manganese(II) oxide, 99.99%, (trace metal basis)
Manganese(II) oxide, 99.99%, (trace metal basis)
Manganese(II) oxide, 99.99%, (trace metal basis)
Acros Organics

Manganese(II) oxide, 99.99%, (trace metal basis)

CAS: 1344-43-0 | MnO | 70.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 436970010
also known as 43697
HSN 28201000
Price (INR)
6,994.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,994.00
Each
Chemical Identifiers
CAS1344-43-0
IUPAC Nameoxomanganese
Molecular FormulaMnO
InChI KeyVASIZKWUTCETSD-UHFFFAOYSA-N
SMILESO=[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Appearance (Color)Green
Titration Complexometric76.7 to 78.2 % (Mn)
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only