Potassium tetrachloroplatinate, 99.99%, (trace metal basis)
Potassium tetrachloroplatinate, 99.99%, (trace metal basis)
Potassium tetrachloroplatinate, 99.99%, (trace metal basis)
Acros Organics

Potassium tetrachloroplatinate, 99.99%, (trace metal basis)

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4370300505 gGlass bottle
Catalog number 437030050
HSN 28439019
Price (INR)
109,788.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
109,788.00
Each
Chemical Identifiers
CAS10025-99-7
IUPAC Namedipotassium tetrachloroplatinumdiuide
Molecular FormulaCl4H4K2Pt
InChI KeyJCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[K+].[K+].[Pt--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Appearance (Form)Crystals
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only