Selenious acid, 99.999%, (trace metal basis)
Selenious acid, 99.999%, (trace metal basis)
Selenious acid, 99.999%, (trace metal basis)
Acros Organics

Selenious acid, 99.999%, (trace metal basis)

CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
437120500
also known as 43712
50 gGlass bottle
Catalog number 437120500
also known as 43712
HSN 28439019
Price (INR)
19,079.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
19,079.00
Each
Chemical Identifiers
CAS7783-00-8
IUPAC Nameselenous acid
Molecular FormulaH2O3Se
InChI KeyMCAHWIHFGHIESP-UHFFFAOYSA-N
SMILESO[Se](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Assay60 to 62.4 % (Se)
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only