Product Image
Applied Biosystems™
MicroSEQ™ ID Fungal Gene Library v2013
Have Questions?
Catalog NumberQuantity
43712961 kit
Catalog number 4371296
Price (INR)
-
Quantity:
1 kit
This separately-available MicroSEQ™ ID Fungal Gene Library v2013 is used to update older versions of the library co-installed with MicroSEQ™ ID Microbial Identification Software . Purchase of this library is not required when purchasing the software, as the most recent library is included with most software purchases. The library contains 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism. The library is regularly updated with new entries and current taxonomy.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntrySequencing run
Data OutputsFungal Identification
For Use With (Equipment)3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
LicenseSeat
Operating SystemWindows Vista, Windows XP (Service Pack 3), Windows 7
Product TypeFungal Gene Library v2013
Quantity1 kit
Software CategorySecondary Analysis
Product LineMicroSEQ
Software TypeIdentification
Unit SizeEach