TaqMan™ RNase P Instrument Verification Plate, Fast 48-well
Product Image
Applied Biosystems™
TaqMan™ RNase P Instrument Verification Plate, Fast 48-well
Applied Biosystems® TaqMan® RNase P Fast 48-Well Instrument Verification Plate is used to verify the performance of the Applied Biosystems®Read more
Have Questions?
Catalog NumberQuantity
43714391 x 48-well plate
Catalog number 4371439
Price (INR)
79,767.00
Each
Add to cart
Quantity:
1 x 48-well plate
Price (INR)
79,767.00
Each
Add to cart
Applied Biosystems® TaqMan® RNase P Fast 48-Well Instrument Verification Plate is used to verify the performance of the Applied Biosystems® StepOne™ Real-Time PCR System. The TaqMan® RNase P Fast 48-Well Instrument Verification Plate is a 48-well PCR plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan® Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The TaqMan® RNase P Fast 48-Well Instrument Verification Plate satisfies the same requirements as the Applied Biosystems® StepOne™ Real-Time PCR Systems installation. To pass installation, the instrument must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems® recommends that the performance of your instrument be verified every six months.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeFAM
Calibration TypeRNase P Calibration, Instrument Verification Plate
Detection MethodFluorescence
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)StepOne™, Fast Mode
Product LineTaqMan
Quantity1 x 48-well plate
Shipping ConditionDry Ice
Product TypeInstrument Verification Plate
Unit SizeEach
Contents & Storage
Contains 48-Well Fast Reaction Plate preloaded and sealed with TaqMan™ RNase P probes and primers. Store at -15 to 25°C. This product is light sensitive.
Frequently asked questions (FAQs)
Can I use my 7900HT System RNase P instrument verification plates with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems?
Only the TaqMan RNase P Instrument Verification Plate, Fast 96-well (for 0.1 mL block) (Cat. No. 4351979) is compatible with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems. The following RNase P instrument verification plates can be used on the 7900HT System, but not on the QuantStudio 6 and 7 Flex Real-Time PCR Systems:
• TaqMan RNase P Instrument Verification Plate, 96-well (Cat. No. 4310982) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for 7900HT System, 384-well (Cat. No. 4323306) (for 384-well block)
• 7900HT TaqMan Low Density Array Instrument Verification RNase P Kit (Cat. No. 4351468) (for TaqMan Array Card block)
The following RNase P instrument verification plates can be used on the QuantStudio 6 and 7 Flex Real-Time PCR Systems, but not on the 7900HT System:
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 96-well (Cat. No. 4432382) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 384-well (Cat. No. 4455280) (for 384-well block)
• ViiA7 Array Card RNase P Verification Kit (Cat. No. 4432265) (for TaqMan Array Card block)
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.