2,6-Pyridinedicarboxaldehyde, 97%
2,6-Pyridinedicarboxaldehyde, 97%
2,6-Pyridinedicarboxaldehyde, 97%
Acros Organics

2,6-Pyridinedicarboxaldehyde, 97%

CAS: 5431-44-7 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 437750010
HSN 29333919
Price (INR)
42,265.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
42,265.00
Each
Chemical Identifiers
CAS5431-44-7
IUPAC Namepyridine-2,6-dicarbaldehyde
Molecular FormulaC7H5NO2
InChI KeyPMWXGSWIOOVHEQ-UHFFFAOYSA-N
SMILESO=CC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Powder
Infrared spectrumConforms
Melting point122°C to 126°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only