Product Image
Applied Biosystems™

TaqMan™ Array Rat Inflammation Panel

The Applied Biosystems™ TaqMan™ Rat Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.
Have Questions?
Catalog NumberQuantity
43787084 cards
Catalog number 4378708
Price (INR)
236,128.00
Each
Quantity:
4 cards
Price (INR)
236,128.00
Each

The Applied Biosystems™ TaqMan™ Rat Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and the panel is sold in packs of 4.

The TaqMan™ Gene Signature Arrays for rat inflammation contain over 90 genes that are, or have been, studied as targets for a range of inflammatory diseases. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryInflammation, Signal Transduction
SpeciesRat
TargetOver 90 genes that are targets for a range of inflammatory diseases
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.