TaqMan™ Array Human Angiogenesis Panel
Applied Biosystems™

TaqMan™ Array Human Angiogenesis Panel

The Applied Biosystems™ TaqMan™ Human Angiogenesis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of angiogenesis genes important in drug discovery.
Have Questions?
Catalog NumberQuantity
43787104 Cards
Catalog number 4378710
Price (INR)
242,908.00
Each
Quantity:
4 Cards
Price (INR)
242,908.00
Each

The Applied Biosystems™ TaqMan™ Human Angiogenesis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of angiogenesis genes important in drug discovery. This product is designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and is sold in packs of 4.

This TaqMan™ Gene Signature Array contains 94 assays and two controls to study angiogenesis and lymphangiogenesis in humans. It includes known angiogenesis growth factors like VEGF, and known matrix derived inhibitors such as endostatin. Additionally, the panel contains markers and targets for angiogenesis and lymphangiogenesis. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 Cards
Research CategoryAngiogenesis, Oncology
SpeciesHuman
TargetVEGF, endostatin, and other targets for angiogenesis and lymphangiogenesis
For Use With (Application)Oncology, Genetic Disease Research
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.