Product Image
Applied Biosystems™

TaqMan™ Array Human Nuclear Receptors Panel

The Applied Biosystems™ TaqMan™ Human Nuclear Receptor Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.
Have Questions?
Catalog NumberQuantity
43799614 cards
Catalog number 4379961
Price (INR)
242,908.00
Each
Quantity:
4 cards
Price (INR)
242,908.00
Each

The Applied Biosystems™ TaqMan™ Human Nuclear Receptor Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 64 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.

The nuclear hormone receptor family is highly conserved in vertebrates. They have a modular domain structure consisting of a DNA-binding domain and a ligand-binding domain. Other domains include a modulatory A⁄B domain, the hinge D domain, and a variable C-terminal F domain that is missing in some nuclear hormone receptors. The TaqMan™ Nuclear Receptor Gene Signature Array contains 48 human nuclear hormone receptors (and 16 controls) and has been classified according to sequence homology into seven subfamilies. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryCell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Development
SpeciesHuman
Target48 human nuclear hormone receptors
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.